- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028097.3 | NP_082373.1 | 7 | 1027 | ||
GenBank mRNA | AK008493.1 | - | 7 | 1061 | ||
AK048210.1 | - | 7 | 1027 | |||
AK084881.1 | - | 7 | 1061 | |||
AK088600.1 | - | 8 | 1197 | |||
AK147765.1 | - | 7 | 1060 | |||
AK162563.1 | - | 7 | 1060 | |||
AK165916.1 | - | 7 | 1089 | |||
AK166237.1 | - | 7 | 1113 | |||
BC016240.1 | - | 7 | 1045 | 4506688 |