Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028097.3 | NP_082373.1 | 7, 8 | 1077 | ||
GenBank mRNA | AK008493.1 | - | 7, 8 | 1111 | ||
AK048210.1 | - | 7, 8 | 1077 | |||
AK084881.1 | - | 7, 8 | 1111 | |||
AK088600.1 | - | 8, 9 | 1247 | |||
AK147765.1 | - | 7, 8 | 1110 | |||
AK162563.1 | - | 7, 8 | 1110 | |||
AK165916.1 | - | 7, 8 | 1139 | |||
AK166237.1 | - | 7, 8 | 1163 | |||
BC016240.1 | - | 7, 8 | 1095 | 4506688 |