Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028108.3 | NP_082384.1 | 3, 4 | 550 | ||
GenBank mRNA | AK011160.1 | - | 3, 4 | 476 | ||
AK135658.1 | - | 3, 4 | 493 | |||
AK145345.1 | - | 3, 4 | 478 | |||
AK156791.1 | - | 3, 4 | 496 | |||
AK167141.1 | - | 4, 5 | 670 | |||
AK168394.1 | - | 3, 4 | 521 | |||
AK184762.1 | - | 2, 3 | 246 | |||
AK195449.1 | - | 2, 3 | 246 | |||
AK198408.1 | - | 3, 4 | 319 | |||
AK219650.1 | - | 2, 3 | 246 | |||
BC004086.1 | - | 3, 4 | 462 | |||
BC055846.1 | - | 3, 4 | 315 | 3600545 | ||
BC057117.1 | - | 3, 4 | 509 | 6409354 |