Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9065
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
7, 8
796
NM_001018017.2
NP_001018017.1
7, 8
769
NM_001044390.2
NP_001037855.1
6, 7
613
NM_001044391.2
NP_001037856.1
4, 5
454
NM_001044392.2
NP_001037857.1
4, 5
481
NM_001044393.2
NP_001037858.1
6, 7
581
NM_001204285.1
NP_001191214.1
7, 8
1429
NM_001204286.1
NP_001191215.1
7, 8
1456
NM_001204287.1
NP_001191216.1
7, 8
850
NM_001204288.1
NP_001191217.1
7, 8
764
NM_001204289.1
NP_001191218.1
6, 7
718
NM_001204290.1
NP_001191219.1
6, 7
655
NM_001204291.1
NP_001191220.1
6, 7
727
NM_001204292.1
NP_001191221.1
6, 7
721
NM_001204293.1
NP_001191222.1
6, 7
701
NM_001204294.1
NP_001191223.1
6, 7
694
NM_001204295.1
NP_001191224.1
5, 6
571
NM_001204296.1
NP_001191225.1
6, 7
640
NM_001204297.1
NP_001191226.1
4, 5
535
NM_002456.5
NP_002447.4
7, 8
823
GenBank mRNA
AF125525.1
-
7, 8
688
AF348143.1
-
4, 5
460
AK301575.1
-
8, 9
1681
AY327582.1
-
6, 7
565
AY327583.1
-
4, 5
406
AY327584.1
-
4, 5
433
AY327585.1
-
4, 5
427
AY327586.1
-
4, 5
454
AY327587.1
-
7, 8
774
AY327588.1
-
6, 7
669
AY327589.1
-
4, 5
459
AY327590.1
-
5, 6
509
AY327591.1
-
6, 7
564
AY327592.1
-
6, 7
584
AY327596.1
-
8, 9
904
AY327597.1
-
5, 6
443
AY327598.1
-
6, 7
533
AY327599.1
-
5, 6
536
AY327600.1
-
6, 7
552
BC120974.2
-
7, 8
746
40115473
BC120975.2
-
7, 8
773
40115475
EF583642.1
-
7, 8
1099
EF583643.1
-
7, 8
927
EF583644.1
-
7, 8
913
EF583645.1
-
7, 8
873
EF583646.1
-
7, 8
841
EF583647.1
-
7, 8
828
EF583648.1
-
7, 8
755
EF583649.1
-
7, 8
742
EF583650.1
-
7, 8
711
EF583651.1
-
7, 8
706
EF583652.1
-
6, 7
696
EF583653.1
-
6, 7
654
EF583654.1
-
6, 7
633
EF583655.1
-
5, 6
581
EF583656.1
-
4, 5
442
EF670706.1
-
7, 8
1123
EF670707.1
-
7, 8
792
EF670708.1
-
6, 7
705
EF670709.1
-
6, 7
699
EF670710.1
-
6, 7
691
EF670711.1
-
6, 7
679
EF670712.1
-
6, 7
672
EF670713.1
-
6, 7
637
EF670714.1
-
6, 7
627
EF670715.1
-
4, 5
621
EF670716.1
-
5, 6
549
EF670718.1
-
4, 5
513
EF670719.1
-
4, 5
483
EF670720.1
-
3, 4
443
EF670721.1
-
4, 5
440
EF670722.1
-
4, 5
417
FJ226029.1
-
6, 7
1388
FJ226031.1
-
6, 7
1316
FJ226032.1
-
6, 7
1249
FJ226033.1
-
6, 7
1230
FJ226034.1
-
7, 8
1149
FJ226036.1
-
5, 6
1112
FJ226038.1
-
4, 5
1055
FJ226041.1
-
8, 9
931
FJ226043.1
-
7, 8
814
FJ226044.1
-
6, 7
798
FJ226045.1
-
6, 7
768
FJ226046.1
-
6, 7
763
FJ226047.1
-
7, 8
728
FJ226049.1
-
6, 7
691
FJ226050.1
-
6, 7
664
FJ226051.1
-
6, 7
637
FJ226052.1
-
6, 7
625
FJ226053.1
-
6, 7
618
FJ226054.1
-
5, 6
614
FJ226055.1
-
5, 6
562
J05581.1
-
7, 8
1429
J05582.1
-
3770
M34088.1
-
5, 6
916
M34089.1
-
5, 6
916
U60259.1
-
6, 7
647
U60260.1
-
7, 8
722
X52229.1
-
8, 9
1534
X80761.1
-
7, 8
697
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline