Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adpgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028121.2 | NP_082397.1 | 6, 7 | 993 | ||
GenBank mRNA | AK011434.1 | - | 6, 7 | 958 | ||
AK162169.1 | - | 6, 7 | 946 | |||
AK169590.1 | - | 6, 7 | 957 |