Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9066
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
7
705
NM_001018017.2
NP_001018017.1
7
678
NM_001044390.2
NP_001037855.1
6
522
NM_001044391.2
NP_001037856.1
4
363
NM_001044392.2
NP_001037857.1
4
390
NM_001044393.2
NP_001037858.1
6
490
NM_001204285.1
NP_001191214.1
7
1338
NM_001204286.1
NP_001191215.1
7
1365
NM_001204287.1
NP_001191216.1
7
759
NM_001204288.1
NP_001191217.1
7
673
NM_001204289.1
NP_001191218.1
6
627
NM_001204290.1
NP_001191219.1
6
564
NM_001204291.1
NP_001191220.1
6
636
NM_001204292.1
NP_001191221.1
6
630
NM_001204293.1
NP_001191222.1
6
610
NM_001204294.1
NP_001191223.1
6
603
NM_001204295.1
NP_001191224.1
5
480
NM_001204296.1
NP_001191225.1
6
549
NM_001204297.1
NP_001191226.1
4
444
NM_002456.5
NP_002447.4
7
732
GenBank mRNA
AF125525.1
-
7
597
AF348143.1
-
4
369
AK301575.1
-
8
1590
AY327582.1
-
6
474
AY327583.1
-
4
315
AY327584.1
-
4
342
AY327585.1
-
4
336
AY327586.1
-
4
363
AY327587.1
-
7
683
AY327588.1
-
6
578
AY327589.1
-
4
368
AY327590.1
-
5
418
AY327591.1
-
6
473
AY327592.1
-
6
493
AY327595.1
-
5
405
AY327596.1
-
8
813
AY327597.1
-
5
352
AY327598.1
-
6
442
AY327599.1
-
5
445
AY327600.1
-
6
461
BC120974.2
-
7
655
40115473
BC120975.2
-
7
682
40115475
EF583642.1
-
7
1008
EF583643.1
-
7
836
EF583644.1
-
7
822
EF583645.1
-
7
782
EF583646.1
-
7
750
EF583647.1
-
7
737
EF583648.1
-
7
664
EF583649.1
-
7
651
EF583650.1
-
7
620
EF583651.1
-
7
615
EF583652.1
-
6
605
EF583653.1
-
6
563
EF583654.1
-
6
542
EF583655.1
-
5
490
EF583656.1
-
4
351
EF670706.1
-
7
1032
EF670707.1
-
7
701
EF670708.1
-
6
614
EF670709.1
-
6
608
EF670710.1
-
6
600
EF670711.1
-
6
588
EF670712.1
-
6
581
EF670713.1
-
6
546
EF670714.1
-
6
536
EF670715.1
-
4
530
EF670716.1
-
5
458
EF670717.1
-
4
368
EF670718.1
-
4
422
EF670719.1
-
4
392
EF670720.1
-
3
352
EF670721.1
-
4
349
EF670722.1
-
4
326
FJ226028.1
-
6
1139
FJ226029.1
-
6
1297
FJ226030.1
-
7
1267
FJ226031.1
-
6
1225
FJ226032.1
-
6
1158
FJ226033.1
-
6
1139
FJ226034.1
-
7
1058
FJ226035.1
-
6
546
FJ226036.1
-
5
1021
FJ226037.1
-
7
836
FJ226038.1
-
4
964
FJ226039.1
-
7
763
FJ226040.1
-
7
683
FJ226041.1
-
8
840
FJ226042.1
-
7
737
FJ226043.1
-
7
723
FJ226044.1
-
6
707
FJ226045.1
-
6
677
FJ226046.1
-
6
672
FJ226047.1
-
7
637
FJ226049.1
-
6
600
FJ226050.1
-
6
573
FJ226051.1
-
6
546
FJ226052.1
-
6
534
FJ226053.1
-
6
527
FJ226054.1
-
5
523
J05581.1
-
7
1338
J05582.1
-
3679
M34088.1
-
5
825
M34089.1
-
5
825
U60259.1
-
6
556
U60260.1
-
7
631
U60261.1
-
7
685
X52228.1
-
4
1916
X52229.1
-
8
1443
X80761.1
-
7
606
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0