Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9067
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
2
245
NM_001018017.2
NP_001018017.1
2
218
NM_001044390.2
NP_001037855.1
2
218
NM_001044391.2
NP_001037856.1
2
218
NM_001044392.2
NP_001037857.1
2
245
NM_001044393.2
NP_001037858.1
2
218
NM_001204285.1
NP_001191214.1
2
218
NM_001204286.1
NP_001191215.1
2
245
NM_001204287.1
NP_001191216.1
2
245
NM_001204288.1
NP_001191217.1
2
245
NM_001204289.1
NP_001191218.1
2
245
NM_001204290.1
NP_001191219.1
2
182
NM_001204291.1
NP_001191220.1
2
245
NM_001204292.1
NP_001191221.1
2
245
NM_001204293.1
NP_001191222.1
2
218
NM_001204294.1
NP_001191223.1
2
218
NM_001204295.1
NP_001191224.1
2
245
NM_001204296.1
NP_001191225.1
2
245
NM_001204297.1
NP_001191226.1
2
245
NM_002456.5
NP_002447.4
2
218
GenBank mRNA
AF125525.1
-
2
137
AF348143.1
-
2
170
AK301575.1
-
2
212
AY327582.1
-
2
170
AY327583.1
-
2
170
AY327584.1
-
2
197
AY327585.1
-
2
170
AY327586.1
-
2
197
AY327587.1
-
2
223
AY327588.1
-
2
196
AY327589.1
-
2
170
AY327590.1
-
2
170
AY327591.1
-
2
170
AY327592.1
-
2
170
AY327595.1
-
2
170
AY327596.1
-
2
196
AY327597.1
-
2
170
AY327599.1
-
2
197
AY327600.1
-
2
170
AY466157.1
-
2
146
BC120974.2
-
2
195
40115473
BC120975.2
-
2
222
40115475
EF583642.1
-
2
223
EF583643.1
-
2
322
EF583644.1
-
2
223
EF583645.1
-
2
322
EF583647.1
-
2
223
EF583648.1
-
2
223
EF583649.1
-
2
223
EF583650.1
-
2
160
EF583651.1
-
2
160
EF583652.1
-
2
223
EF583653.1
-
2
223
EF583654.1
-
2
160
EF583655.1
-
2
223
EF583656.1
-
2
223
EF670706.1
-
2
187
EF670707.1
-
2
187
EF670708.1
-
2
223
EF670709.1
-
2
223
EF670710.1
-
2
196
EF670711.1
-
2
196
EF670712.1
-
2
196
EF670713.1
-
2
223
EF670714.1
-
2
196
EF670715.1
-
2
187
EF670716.1
-
2
223
EF670717.1
-
2
223
EF670718.1
-
2
223
EF670719.1
-
2
223
EF670720.1
-
2
160
EF670721.1
-
2
223
EF670722.1
-
2
160
EF670723.1
-
2
196
FJ226028.1
-
1
698
FJ226029.1
-
1
698
FJ226030.1
-
2
668
FJ226031.1
-
1
698
FJ226032.1
-
1
698
FJ226033.1
-
1
698
FJ226035.1
-
2
223
FJ226036.1
-
1
698
FJ226037.1
-
2
322
FJ226038.1
-
1
697
FJ226039.1
-
2
322
FJ226040.1
-
2
223
FJ226041.1
-
2
223
FJ226042.1
-
2
223
FJ226043.1
-
2
196
FJ226044.1
-
2
322
FJ226045.1
-
2
295
FJ226046.1
-
2
295
FJ226047.1
-
2
196
FJ226049.1
-
2
223
FJ226050.1
-
2
196
FJ226052.1
-
2
196
FJ226053.1
-
2
223
FJ226054.1
-
2
322
FJ226055.1
-
2
223
J05581.1
-
2
218
J05582.1
-
219
M31823.1
-
2
239
M32738.1
-
2
223
M32739.1
-
2
196
S81736.1
-
2
171
S81781.1
-
2
182
U60259.1
-
2
171
U60260.1
-
2
171
U60261.1
-
2
171
X52228.1
-
2
203
X52229.1
-
2
203
X80761.1
-
2
146
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0