Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291070.1 | NP_001277999.1 | 16 | 1677 | ||
NM_175178.4 | NP_780387.2 | 18 | 2083 | |||
GenBank mRNA | AK049928.1 | - | 17 | 3070 | ||
AK158809.1 | - | 17 | 1745 | |||
BC046517.1 | - | 18 | 2070 | |||
BC096476.1 | - | 17 | 1858 | 4221224 |