Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028198.2 | NP_082474.1 | 12 | 1342 | ||
GenBank mRNA | AF343581.1 | - | 13 | 1342 | ||
AK010389.1 | - | 12 | 1373 | |||
BC039187.1 | - | 269 | ||||
BC092371.1 | - | 1415 | ||||
BC131660.1 | - | 12 | 1338 | 40110670 | ||
BC131661.1 | - | 12 | 1338 | 40110672 |