Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028203.1 | NP_082479.1 | 2 | 186 | ||
GenBank mRNA | AK011125.1 | - | 3 | 388 | ||
AK132326.1 | - | 2 | 157 | |||
BC111903.1 | - | 2 | 151 | 40062287 | ||
BC112430.1 | - | 2 | 151 | 40062286 |