Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUTYH SIRNA Assays ›
Gene Symbol:
MUTYH
Gene Name:
mutY DNA glycosylase
Gene Aliases:
MYH
Chromosome Location:
Chr.1: 45329242 - 45340925 on Build GRCh38
UniGene ID:
Hs.271353
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9092
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUTYH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048171.1
NP_001041636.1
14
1529
NM_001048172.1
NP_001041637.1
14
1420
NM_001048173.1
NP_001041638.1
14
1417
NM_001048174.1
NP_001041639.1
14
1336
NM_001128425.1
NP_001121897.1
14
1571
NM_001293190.1
NP_001280119.1
14
1532
NM_001293191.1
NP_001280120.1
14
1369
NM_001293192.1
NP_001280121.1
14
1465
NM_001293195.1
NP_001280124.1
15
1486
NM_001293196.1
NP_001280125.1
14
1354
NM_012222.2
NP_036354.1
14
1562
XM_011541497.2
XP_011539799.1
14
1628
XM_011541498.1
XP_011539800.1
14
1406
XM_011541499.1
XP_011539801.1
14
1392
XM_011541500.2
XP_011539802.1
14
1587
XM_011541501.2
XP_011539803.1
15
1501
XM_011541502.2
XP_011539804.1
14
1406
XM_011541503.2
XP_011539805.1
13
1374
XM_011541504.2
XP_011539806.1
14
1455
XM_011541505.2
XP_011539807.1
10
1075
XM_011541506.1
XP_011539808.1
10
1059
XM_011541507.2
XP_011539809.2
9
871
XM_017001331.1
XP_016856820.1
14
1405
XM_017001332.1
XP_016856821.1
14
1407
XM_017001333.1
XP_016856822.1
14
1460
XM_017001334.1
XP_016856823.1
14
1353
XM_017001335.1
XP_016856824.1
14
1299
XM_017001336.1
XP_016856825.1
13
1383
XM_017001337.1
XP_016856826.1
13
1323
XM_017001338.1
XP_016856827.1
13
1910
XM_017001339.1
XP_016856828.1
13
1361
GenBank mRNA
AB032920.1
-
14
1480
AB032921.1
-
14
1450
AB032922.1
-
14
1447
AB032923.1
-
14
1383
AB032924.1
-
14
1347
AB032925.1
-
14
1314
AB032926.1
-
15
1464
AB032927.1
-
14
1385
AB032928.1
-
14
1382
AB032929.1
-
14
1318
AK293830.1
-
5
591
BC003178.1
-
14
1467
2958002
KU178203.1
-
14
1313
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA glycosylase
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
mismatch repair
response to oxidative stress
depurination
DNA binding
protein binding
DNA N-glycosylase activity
MutLalpha complex binding
MutLbeta complex binding
MutSalpha complex binding
MutSbeta complex binding
metal ion binding
4 iron, 4 sulfur cluster binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0