Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028228.3 | NP_082504.1 | 2 | 194 | ||
GenBank mRNA | AF321817.1 | - | 2 | 142 | ||
AF421879.1 | - | 2 | 77 | |||
AK012057.1 | - | 2 | 172 | |||
BC065781.1 | - | 2 | 93 | |||
BC115636.1 | - | 2 | 106 | 40102952 | ||
BC115637.1 | - | 2 | 104 |