Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028232.2 | NP_082508.1 | 7 | 742 | ||
GenBank mRNA | AB193067.1 | - | 6 | 650 | ||
AB193068.1 | - | 6 | 650 | |||
AK014357.1 | - | 7 | 736 | |||
AK049517.1 | - | 3 | 106 | |||
AK154188.1 | - | 7 | 761 | |||
BC089014.1 | - | 6 | 589 | 30604765 |