Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289591.1 | NP_001276520.1 | 3 | 625 | ||
NM_001289592.1 | NP_001276521.1 | 4 | 592 | |||
NM_028261.1 | NP_082537.1 | 4 | 715 | |||
GenBank mRNA | AK012006.1 | - | 4 | 662 | ||
AK077788.1 | - | 4 | 616 | |||
AK089405.1 | - | 4 | 693 | |||
AK146284.1 | - | 4 | 588 | |||
AK153868.1 | - | 4 | 586 | |||
AK157370.1 | - | 3 | 527 | |||
AK158458.1 | - | 4 | 501 | |||
AK170724.1 | - | 4 | 693 | |||
AK171065.1 | - | 4 | 679 | |||
AK171203.1 | - | 4 | 679 | |||
AK171612.1 | - | 4 | 646 | |||
AK172314.1 | - | 3 | 457 | |||
AX775911.1 | - | 4 | 602 | |||
BC027757.1 | - | 2 | 359 | |||
BC046640.1 | - | 3 | 550 | 5067343 | ||
DQ910493.1 | - | 4 | 513 | |||
FJ222242.1 | - | 2 | 377 |