Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYB SIRNA Assays ›
Gene Symbol:
MYB
Gene Name:
MYB proto-oncogene, transcription factor
Gene Aliases:
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location:
Chr.6: 135180981 - 135219173 on Build GRCh38
UniGene ID:
Hs.606320
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9109
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130172.1
NP_001123644.1
8
1080
NM_001130173.1
NP_001123645.1
8
1080
NM_001161656.1
NP_001155128.1
8
1080
NM_001161657.1
NP_001155129.1
8
1080
NM_001161658.1
NP_001155130.1
8
1080
NM_001161659.1
NP_001155131.1
8
1080
NM_005375.3
NP_005366.2
8
1080
NR_134958.1
8
1080
NR_134959.1
8
1080
NR_134960.1
8
1080
NR_134961.1
8
1080
NR_134962.1
7
859
NR_134963.1
8
1080
NR_134964.1
8
1080
NR_134965.1
8
1080
GenBank mRNA
AF104863.1
-
8
958
AJ606317.1
-
8
1080
AJ606318.1
-
8
1080
AJ606319.1
-
8
1080
AJ606320.1
-
8
1080
AJ606321.1
-
8
1080
AJ606322.1
-
8
1080
AJ606323.1
-
8
1080
AJ606324.1
-
8
1080
AJ616235.1
-
8
995
AJ616798.1
-
1
40
AY787443.1
-
8
881
AY787444.1
-
6
588
AY787445.1
-
7
809
AY787446.1
-
7
660
AY787448.1
-
8
881
AY787449.1
-
8
881
AY787450.1
-
8
881
AY787451.1
-
8
881
AY787452.1
-
8
881
AY787453.1
-
8
881
AY787454.1
-
8
881
AY787455.1
-
8
881
AY787456.1
-
8
881
AY787457.1
-
8
881
AY787458.1
-
8
881
AY787459.1
-
8
881
AY787460.1
-
8
881
AY787461.1
-
8
881
AY787462.1
-
8
881
AY787463.1
-
8
881
AY787464.1
-
8
881
AY787465.1
-
8
881
AY787466.1
-
8
881
AY787467.1
-
8
881
AY787468.1
-
8
881
AY787469.1
-
8
881
AY787470.1
-
8
881
AY787471.1
-
8
881
AY787472.1
-
8
881
AY787473.1
-
8
881
AY787474.1
-
8
881
AY787475.1
-
8
881
BC035996.1
-
2
120
BC064955.1
-
8
1032
6069320
M13665.1
-
6
743
M13666.1
-
6
743
M15024.1
-
8
994
X52125.1
-
8
1078
X52126.1
-
7
884
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008758639.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
chromatin remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of T-helper cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-b8dk7:80/10.239.160.22:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0