Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028295.1 | NP_082571.1 | 14 | 1334 | ||
GenBank mRNA | AK012415.1 | - | 2 | 125 | ||
BC009151.1 | - | 14 | 1334 | 3600849 |