Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144802.4 | NP_659051.3 | 3 | 953 | ||
GenBank mRNA | AK012866.1 | - | 3 | 669 | ||
AK034860.1 | - | 3 | 898 | |||
AK048900.1 | - | 3 | 742 | |||
AK146879.1 | - | 3 | 915 | |||
AK160943.1 | - | 2 | 761 | |||
AK171091.1 | - | 3 | 820 | |||
BC012849.1 | - | 3 | 899 | 3587716 |