Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204152.1 | NP_001191081.1 | 3, 4 | 428 | ||
NM_028331.5 | NP_082607.3 | 3 | 594 | |||
GenBank mRNA | AK012868.1 | - | 3, 4 | 370 | ||
AK051058.1 | - | 3 | 508 | |||
AK154407.1 | - | 3 | 571 | |||
BC071187.1 | - | 3 | 561 | 6395221 | ||
DQ002399.1 | - | 3 | 560 |