Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem161b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175187.5 | NP_780396.2 | 2 | 204 | ||
GenBank mRNA | AK035319.1 | - | 2 | 191 | ||
AK088393.1 | - | 2 | 176 | |||
BC051663.1 | - | 2 | 152 | 30020101 | ||
BC060067.1 | - | 2 | 186 | |||
BY239899.1 | - | 2 | 204 |