Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112722.1 | NP_001106193.1 | 12 | 1289 | ||
NM_001112723.1 | NP_001106194.1 | 12 | 1262 | |||
NM_001290803.1 | NP_001277732.1 | 13 | 1388 | |||
NM_172415.3 | NP_766003.3 | 13 | 1379 | |||
GenBank mRNA | AK028570.1 | - | 6 | 704 | ||
AK028648.1 | - | 12 | 1272 | |||
AK129408.1 | - | 12 | 1262 | |||
BC034061.1 | - | 6 | 525 | |||
BC059220.1 | - | 13 | 1367 | 6417641 | ||
CX564261.1 | - | 2 | 181 |