Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 3 | 480 | ||
GenBank mRNA | AF512670.1 | - | 3 | 186 | ||
AF536771.1 | - | 3 | 249 | |||
AK013600.1 | - | 3 | 482 | |||
AK018119.1 | - | 3 | 437 | |||
AK039592.1 | - | 3 | 323 | |||
AK079059.1 | - | 930 | ||||
AK086561.1 | - | 1 | 666 | |||
AK148956.1 | - | 3 | 451 | |||
BC026686.1 | - | 3 | 418 | 5065516 | ||
BC066007.1 | - | 3 | 720 | |||
BC085147.1 | - | 3 | 632 | |||
BC088979.1 | - | 3 | 415 | 5690600 |