Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 9 | 1355 | ||
GenBank mRNA | AF512670.1 | - | 9 | 1061 | ||
AF536771.1 | - | 9 | 1124 | |||
AK013600.1 | - | 9 | 1357 | |||
AK018119.1 | - | 9 | 1318 | |||
AK039592.1 | - | 9 | 1198 | |||
AK148956.1 | - | 9 | 1326 | |||
BC026686.1 | - | 9 | 1293 | 5065516 | ||
BC066007.1 | - | 9 | 1596 | |||
BC085147.1 | - | 9 | 1508 | |||
BC088979.1 | - | 9 | 1290 | 5690600 |