Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 5 | 844 | ||
GenBank mRNA | AF512670.1 | - | 5 | 550 | ||
AF536771.1 | - | 5 | 613 | |||
AK013600.1 | - | 5 | 846 | |||
AK018119.1 | - | 5 | 801 | |||
AK039592.1 | - | 5 | 687 | |||
AK079059.1 | - | 1294 | ||||
AK148956.1 | - | 5 | 815 | |||
BC026686.1 | - | 5 | 782 | 5065516 | ||
BC066007.1 | - | 5 | 1084 | |||
BC085147.1 | - | 5 | 997 | |||
BC088979.1 | - | 5 | 779 | 5690600 |