- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028416.2 | NP_082692.1 | 3 | 352 | ||
GenBank mRNA | AJ457192.1 | - | 3 | 364 | ||
BC120532.1 | - | 3 | 295 | 40129455 | ||
BC125438.1 | - | 3 | 295 | 40129953 |