Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160043.1 | NP_001153515.1 | 2 | 868 | ||
NM_028457.2 | NP_082733.1 | 2 | 889 | |||
GenBank mRNA | AK014134.1 | - | 2 | 887 | ||
BC023360.1 | - | 2 | 882 | 5064775 |