Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028499.2 | NP_082775.1 | 4 | 422 | ||
GenBank mRNA | AK006601.1 | - | 4 | 433 | ||
AK161371.1 | - | 4 | 425 |