Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_079420.2 | NP_524144.1 | 3 | 309 | ||
NM_079422.2 | NP_524146.1 | 3 | 119 | |||
GenBank mRNA | AK311892.1 | - | 3 | 119 | ||
AK311942.1 | - | 3 | 330 | |||
BC005318.1 | - | 3 | 119 | 3933998 | ||
CR456869.1 | - | 3 | 180 | |||
DA898482.1 | - | 3 | 335 |