Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speer4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028561.2 | NP_082837.1 | 5 | 1764 | ||
GenBank mRNA | AK006975.1 | - | 5 | 1813 |
Gene Symbol: Gm10220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134299.1 | NP_001127771.1 | 5 | 1816 |
Gene Symbol: Gm10471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177579.1 | NP_001171050.1 | 5 | 1758 |