Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cybrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028593.2 | NP_082869.2 | 4 | 782 | ||
GenBank mRNA | AK008849.1 | - | 4 | 728 | ||
AK029112.1 | - | 4 | 785 | |||
BC027665.1 | - | 2 | 278 | |||
BC147289.1 | - | 2 | 270 | |||
BC147290.1 | - | 2 | 270 |