Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028665.4 | NP_082941.2 | 5 | 1419 | ||
GenBank mRNA | AK029511.1 | - | 5 | 1421 | ||
AK029888.1 | - | 5 | 1380 | |||
AK033599.1 | - | 5 | 1408 | |||
AK048925.1 | - | 5 | 1313 | |||
AK133339.1 | - | 1380 | ||||
BC030885.1 | - | 5 | 1048 | |||
BC100433.1 | - | 5 | 1455 | |||
BC145456.1 | - | 5 | 537 | 9054017 |