Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025947.2 | NP_001021118.1 | 13 | 1636 | ||
NM_001276340.1 | NP_001263269.1 | 13 | 1654 | |||
NM_001276341.1 | NP_001263270.1 | 12 | 1398 | |||
NM_152816.3 | NP_690029.2 | 14 | 1675 | |||
GenBank mRNA | AB079133.1 | - | 13 | 1488 | ||
AK018195.1 | - | 14 | 1615 | |||
AK051443.1 | - | 13 | 1565 | |||
AK080871.1 | - | 13 | 1556 | |||
AK154123.1 | - | 13 | 1405 | |||
AK190489.1 | - | 3 | 216 | |||
BC040777.1 | - | 12 | 1368 | 4948368 | ||
BC079635.1 | - | 13 | 1485 | 6400313 | ||
BC085158.1 | - | 14 | 1580 | |||
BQ550714.1 | - | 5 | 572 | |||
CD559921.1 | - | 5 | 498 | |||
CK341370.1 | - | 3 | 149 | |||
CK391623.1 | - | 5 | 423 | |||
DV652251.1 | - | 6 | 663 |