Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080527.1 | NP_001073996.1 | 43, 44 | 5988 | ||
XM_006712539.3 | XP_006712602.1 | 44, 45 | 6179 | |||
XM_011511218.2 | XP_011509520.1 | 44, 45 | 6188 | |||
XM_017004169.1 | XP_016859658.1 | 43, 44 | 6245 | |||
GenBank mRNA | AB290182.1 | - | 19, 20 | 2842 | ||
AK074183.1 | - | 17, 18 | 3212 | |||
BC035615.2 | - | 12, 13 | 1600 | |||
CR627350.1 | - | 1480 |