Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028724.4 | NP_083000.5 | 11 | 2392 | ||
GenBank mRNA | AK014548.1 | - | 3 | 864 | ||
AK040763.1 | - | 3 | 864 | |||
AK150680.1 | - | 9 | 2200 | |||
AK151332.1 | - | 9 | 2156 | |||
AK152025.1 | - | 4 | 837 | |||
AK152893.1 | - | 9 | 2200 | |||
AK154421.1 | - | 3 | 808 | |||
AK163047.1 | - | 3 | 1379 | |||
AK171064.1 | - | 3 | 779 | |||
AK198881.1 | - | 2 | 179 | |||
BC005529.1 | - | 3 | 825 | 3495681 | ||
BC040390.1 | - | 11 | 2386 | 3156682 |