Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632428N05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159572.1 | NP_001153044.1 | 2 | 610 | ||
NM_028732.4 | NP_083008.1 | 2 | 610 | |||
GenBank mRNA | AK014600.1 | - | 2 | 538 | ||
AK030479.1 | - | 2 | 550 | |||
AK032383.1 | - | 2 | 555 | |||
AK033393.1 | - | 2 | 556 | |||
AK079734.1 | - | 2 | 558 | |||
AK131118.1 | - | 2 | 526 | |||
AK159086.1 | - | 2 | 555 | |||
AK159864.1 | - | 2 | 555 | |||
AK162635.1 | - | 449 | ||||
AK163118.1 | - | 2 | 555 | |||
AK170059.1 | - | 2 | 558 | |||
AK170419.1 | - | 2 | 555 | |||
BC003967.1 | - | 2 | 526 | 3488482 | ||
EF426799.1 | - | 2 | 439 | |||
JN602184.1 | - | 2 | 439 |