Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144533.2 | NP_653116.1 | 3 | 577 | ||
GenBank mRNA | AK041526.1 | - | 3 | 306 | ||
AK041897.1 | - | 2 | 352 | |||
BC005737.1 | - | 3 | 577 | 3599134 | ||
BC092086.1 | - | 3 | 286 | 5040019 |