Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144533.2 | NP_653116.1 | 2 | 531 | ||
GenBank mRNA | AK016744.1 | - | 2 | 256 | ||
AK040783.1 | - | 2 | 512 | |||
AK041526.1 | - | 2 | 260 | |||
BC005737.1 | - | 2 | 531 | 3599134 | ||
BC092086.1 | - | 2 | 240 | 5040019 |