Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028782.2 | NP_083058.2 | 3 | 571 | ||
GenBank mRNA | AK004820.1 | - | 3 | 555 | ||
AK131910.1 | - | 3 | 557 | |||
AK157474.1 | - | 3 | 550 | |||
AK159302.1 | - | 3 | 556 | |||
AK160071.1 | - | 3 | 556 | |||
AK160076.1 | - | 3 | 557 | |||
AK161983.1 | - | 3 | 556 | |||
AK177008.1 | - | 1 | 47 | |||
AY190302.1 | - | 3 | 541 | |||
BC057365.1 | - | 3 | 658 | |||
BC058735.1 | - | 3 | 658 |