Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023738.4 | NP_076227.1 | 9 | 1140 | ||
GenBank mRNA | AF288694.1 | - | 4 | 402 | ||
AK004894.2 | - | 9 | 1141 | |||
AK054410.1 | - | 9 | 1147 | |||
AK089235.1 | - | 9 | 1144 | |||
AK129009.1 | - | 9 | 1052 | |||
AK170421.1 | - | 9 | 1133 | |||
AK170644.1 | - | 8 | 1515 | |||
AK172467.1 | - | 8 | 1478 | |||
BC037693.1 | - | 7 | 914 | |||
BC138843.1 | - | 9 | 1057 | 8861865 |