Search Thermo Fisher Scientific
Gene Symbol: NAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321312.1 | NP_001308241.1 | 2 | 593 | ||
NM_001321313.1 | NP_001308242.1 | 2 | 379 | |||
NM_001321314.1 | NP_001308243.1 | 1 | 302 | |||
NM_001321315.1 | NP_001308244.1 | 1 | 302 | |||
NM_005966.3 | NP_005957.2 | 4 | 855 | |||
XM_005246582.1 | XP_005246639.1 | 3 | 528 | |||
XM_005246583.1 | XP_005246640.1 | 3 | 642 | |||
XM_005246586.1 | XP_005246643.1 | 2 | 335 | |||
XM_005246587.1 | XP_005246644.1 | 2 | 335 | |||
XM_011511219.2 | XP_011509521.1 | 3 | 497 | |||
XM_017004170.1 | XP_016859659.1 | 3 | 538 | |||
XM_017004171.1 | XP_016859660.1 | 2 | 708 | |||
XM_017004172.1 | XP_016859661.1 | 2 | 480 | |||
XM_017004173.1 | XP_016859662.1 | 3 | 563 | |||
XM_017004174.1 | XP_016859663.1 | 2 | 379 | |||
XM_017004175.1 | XP_016859664.1 | 2 | 544 | |||
XM_017004176.1 | XP_016859665.1 | 2 | 447 | |||
GenBank mRNA | AF045451.1 | - | 1 | 302 | ||
AF045452.1 | - | 1 | 302 | |||
AK292446.1 | - | 2 | 393 | |||
AK313542.1 | - | 1 | 400 | |||
BC035724.1 | - | 4 | 744 | 5721972 | ||
U47007.1 | - | 1 | 302 |