Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028812.3 | NP_083088.1 | 3 | 725 | ||
GenBank mRNA | AK011543.1 | - | 3 | 726 | ||
AK031528.1 | - | 3 | 715 | |||
AK080478.1 | - | 3 | 660 | |||
AK154283.1 | - | 3 | 660 | |||
BC056966.1 | - | 3 | 745 | 6817212 |