Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028870.3 | NP_083146.1 | 5 | 487 | ||
GenBank mRNA | AK009844.1 | - | 3 | 148 | ||
AK029382.1 | - | 5 | 488 | |||
AK146816.1 | - | 560 | ||||
AK159480.1 | - | 5 | 537 | |||
AK167531.1 | - | 560 | ||||
AK213693.1 | - | 3 | 170 | |||
BC037505.1 | - | 2 | 122 | |||
BC070404.1 | - | 6 | 543 | 5699653 | ||
BC128349.1 | - | 2 | 113 |