Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028881.2 | NP_083157.1 | 10, 11 | 1133 | ||
GenBank mRNA | AK014553.1 | - | 5, 6 | 1504 | ||
AK156192.1 | - | 10, 11 | 1112 | |||
AK169944.1 | - | 6, 7 | 782 | |||
BC023091.1 | - | 6, 7 | 723 | 4023064 | ||
BC023831.1 | - | 10, 11 | 1128 | 5345301 | ||
BC032183.1 | - | 10, 11 | 1138 | 5310152 | ||
BC033300.1 | - | 10, 11 | 1128 | 4166411 | ||
CV783724.1 | - | 7, 8 | 637 |