Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026665.4 | NP_080941.3 | 9 | 1225 | ||
GenBank mRNA | AK014862.1 | - | 6 | 858 | ||
AK014873.1 | - | 9 | 1172 | |||
AK014982.1 | - | 7 | 683 | |||
AK016484.1 | - | 9 | 1009 | |||
AK019414.1 | - | 9 | 1105 | |||
AK028408.1 | - | 8 | 997 | |||
AK028458.1 | - | 9 | 1053 | |||
AY225093.1 | - | 9 | 1018 | |||
AY251192.1 | - | 9 | 1170 | |||
BC025588.1 | - | 8 | 1005 | |||
BC050785.1 | - | 7 | 736 | 6703205 | ||
BC056968.1 | - | 9 | 1469 |