Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028918.2 | NP_083194.2 | 3, 4 | 565 | ||
GenBank mRNA | AK016657.1 | - | 3, 4 | 496 | ||
AK161541.1 | - | 3, 4 | 504 | |||
AK162855.1 | - | 3, 4 | 505 | |||
BC050805.1 | - | 3, 4 | 589 | 6771545 |