Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028918.2 | NP_083194.2 | 8 | 1250 | ||
GenBank mRNA | AK016657.1 | - | 8 | 1181 | ||
AK136690.1 | - | 2 | 95 | |||
AK161541.1 | - | 8 | 1189 | |||
AK162855.1 | - | 8 | 1190 | |||
BC050805.1 | - | 8 | 1274 | 6771545 |