Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Cfap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028948.2 | NP_083224.2 | 7 | 1360 | ||
GenBank mRNA | AK016821.1 | - | 8 | 1571 | ||
BC099500.1 | - | 7 | 1278 | 30932292 | ||
BC125572.1 | - | 7 | 1250 | 40130087 | ||
BC125574.1 | - | 7 | 1250 | 40130089 |