Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204084.2 | NP_001191013.1 | 2 | 344 | ||
NM_001204085.1 | NP_001191014.1 | 2 | 262 | |||
NM_001204086.2 | NP_001191015.1 | 2 | 371 | |||
NM_001278164.1 | NP_001265093.1 | 2 | 253 | |||
NM_001278165.1 | NP_001265094.1 | 2 | 320 | |||
NM_001278166.1 | NP_001265095.1 | 2 | 164 | |||
NM_005380.7 | NP_005371.2 | 2 | 377 | |||
NM_182744.3 | NP_877421.2 | 2 | 272 | |||
GenBank mRNA | AK292101.1 | - | 2 | 303 | ||
AK300872.1 | - | 2 | 272 | |||
AK309257.1 | - | 2 | 371 | |||
AK313265.1 | - | 2 | 262 | |||
AL703187.1 | - | 2 | 270 | |||
AY049783.1 | - | 1 | 101 | |||
BC012037.1 | - | 2 | 173 | 3874204 | ||
BQ671197.1 | - | 229 | ||||
BT019423.1 | - | 1 | 81 | |||
D28124.1 | - | 2 | 142 | |||
DA384331.1 | - | 2 | 320 | |||
DA921297.1 | - | 2 | 269 | |||
DA924824.1 | - | 2 | 344 | |||
DB016315.1 | - | 2 | 251 | |||
DB253522.1 | - | 2 | 272 | |||
DB482000.1 | - | 2 | 237 | |||
HY084536.1 | - | 2 | 160 |
Gene Symbol: MINOS1-NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204088.1 | NP_001191017.1 | 3 | 421 | ||
NM_001204089.1 | NP_001191018.1 | 2 | 236 | |||
GenBank mRNA | AK289456.1 | - | 2 | 234 | ||
FY212253.1 | - | 3 | 330 |