Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302489.1 | NP_001289418.1 | 5 | 560 | ||
NM_001302490.1 | NP_001289419.1 | 4 | 492 | |||
NM_001610.3 | NP_001601.1 | 5 | 588 | |||
GenBank mRNA | AK298439.1 | - | 5 | 563 | ||
AK299382.1 | - | 5 | 517 | |||
AK300569.1 | - | 4 | 516 | |||
AK301860.1 | - | 4 | 450 | |||
AK308512.1 | - | 3 | 807 | |||
AY007137.1 | - | 2 | 120 | |||
BC003160.1 | - | 5 | 533 | 2957907 | ||
BC093010.1 | - | 5 | 546 | 30332133 | ||
X12548.1 | - | 5 | 542 |