Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028995.3 | NP_083271.1 | 12 | 1440 | ||
GenBank mRNA | AK018640.1 | - | 12 | 1417 | ||
AK049696.1 | - | 12 | 1475 | |||
AK087811.1 | - | 12 | 1443 | |||
AK138843.1 | - | 12 | 1441 | |||
AK148206.1 | - | 12 | 1462 | |||
BC057168.1 | - | 12 | 1380 | 4973200 |