Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gvin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039160.2 | NP_001034249.2 | 2 | 934 | ||
NM_029000.3 | NP_083276.2 | 4 | 1128 | |||
GenBank mRNA | AK154796.1 | - | 4 | 1117 |
Gene Symbol: Gm4070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243039.1 | NP_001229968.1 | 4 | 1125 | ||
NM_001243040.1 | NP_001229969.1 | 2 | 931 | |||
GenBank mRNA | AK036418.1 | - | 4 | 1115 | ||
AY167972.1 | - | 4 | 1114 | |||
AY167973.1 | - | 2 | 904 | |||
AY167974.1 | - | 4 | 1098 |